Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300007004.mm.1 |
NM_001146043.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007004.mm.1 |
NM_001271463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007004.mm.1 |
NM_001271464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007004.mm.1 |
NM_001271465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007004.mm.1 |
NM_177266.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007004.mm.1 |
TC1300001099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006993.mm.1 |
NM_001146043.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006993.mm.1 |
NM_001271463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006993.mm.1 |
NM_001271464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006993.mm.1 |
NM_001271465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006993.mm.1 |
NM_177266.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006993.mm.1 |
TC1300001099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300007005.mm.1 |
NM_001146043.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007005.mm.1 |
NM_001271463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007005.mm.1 |
NM_001271464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007005.mm.1 |
NM_001271465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007005.mm.1 |
NM_177266.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007005.mm.1 |
TC1300001099.mm.1 |
MTA TC |