Assay Details
| Gene Symbol: Hmgcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008255.2 | NP_032281.2 | 12 - 13 | 1705 | 75 | |
| GenBank mRNA | AK159899.1 | - | 11 - 12 | 1593 | 75 | |
| BC019782.1 | - | 11 - 12 | 1581 | 75 | ||
| BC059873.1 | - | 3 - 4 | 386 | 75 | ||
| BC085083.1 | - | 12 - 13 | 1617 | 5320486 | 75 | |
More Information
Set Membership: |
|

Hmgcr
Genomic Map