Assay Details
| Gene Symbol: Hmgcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008255.2 | NP_032281.2 | 17 - 18 | 2443 | 76 | |
| GenBank mRNA | AK159899.1 | - | 16 - 17 | 2331 | 76 | |
| BC019782.1 | - | 16 - 17 | 2319 | 76 | ||
| BC059873.1 | - | 8 - 9 | 1124 | 76 | ||
| BC085083.1 | - | 17 - 18 | 2355 | 5320486 | 76 | |
| M62766.1 | - | 2 - 3 | 305 | 76 | ||
More Information
Set Membership: |
|

Hmgcr
Genomic Map