Assay Details
| Gene Symbol: Polk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012048.2 | NP_036178.1 | 12 - 13 | 1714 | 109 | |
| GenBank mRNA | AB027563.1 | - | 12 - 13 | 1702 | 109 | |
| AF163571.1 | - | 12 - 13 | 1714 | 109 | ||
| AY769920.1 | - | 11 - 12 | 1351 | 109 | ||
| AY769921.1 | - | 10 - 11 | 1288 | 109 | ||
| AY769923.1 | - | 8 - 9 | 1088 | 109 | ||
| AY769924.1 | - | 8 - 9 | 996 | 109 | ||
| BC052820.1 | - | 9 - 10 | 1379 | 30057338 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polk
Genomic Map