| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300016561.mm.1 |
NM_012048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016561.mm.1 |
TC1300002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016557.mm.1 |
NM_012048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016557.mm.1 |
TC1300002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016560.mm.1 |
NM_012048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016560.mm.1 |
TC1300002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016559.mm.1 |
NM_012048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016559.mm.1 |
TC1300002471.mm.1 |
MTA TC |