Assay Details
| Gene Symbol: Polk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012048.2 | NP_036178.1 | 5 - 6 | 723 | 119 | |
| GenBank mRNA | AB027563.1 | - | 5 - 6 | 711 | 119 | |
| AF163571.1 | - | 5 - 6 | 723 | 119 | ||
| AK138299.1 | - | 5 - 6 | 726 | 119 | ||
| AY769919.1 | - | 4 - 5 | 537 | 119 | ||
| AY769920.1 | - | 4 - 5 | 537 | 119 | ||
| AY769921.1 | - | 4 - 5 | 537 | 119 | ||
| AY769922.1 | - | 4 - 5 | 537 | 119 | ||
| AY769923.1 | - | 4 - 5 | 537 | 119 | ||
| AY769924.1 | - | 4 - 5 | 537 | 119 | ||
| AY769925.1 | - | 4 - 5 | 537 | 119 | ||
| AY769926.1 | - | 4 - 5 | 537 | 119 | ||
| AY769927.1 | - | 4 - 5 | 537 | 119 | ||
| AY769928.1 | - | 4 - 5 | 537 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polk
Genomic Map