Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 13 - 14 | 2009 | 65 | |
| GenBank mRNA | AF461761.1 | - | 13 - 14 | 1908 | 65 | |
| AK007424.1 | - | 4 - 5 | 529 | 65 | ||
| AK051183.1 | - | 13 - 14 | 2009 | 65 | ||
| AK088004.1 | - | 12 - 13 | 1753 | 65 | ||
| AK090176.1 | - | 13 - 14 | 1918 | 65 | ||
| AK166006.1 | - | 13 - 14 | 1819 | 65 | ||
| BC029033.1 | - | 10 - 11 | 1451 | 65 | ||
| BC029217.1 | - | 6 - 7 | 861 | 65 | ||
| BC033452.1 | - | 13 - 14 | 1908 | 3156622 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a5
Genomic Map