Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 2 - 3 | 447 | 137 | |
| GenBank mRNA | AF461761.1 | - | 2 - 3 | 346 | 137 | |
| AK051183.1 | - | 2 - 3 | 447 | 137 | ||
| AK088004.1 | - | 2 - 3 | 362 | 137 | ||
| AK090176.1 | - | 2 - 3 | 356 | 137 | ||
| AK166006.1 | - | 2 - 3 | 257 | 137 | ||
| BC033452.1 | - | 2 - 3 | 346 | 3156622 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051183.1, AK088004.1 transcripts.
Set Membership: |
|

Slc30a5
Genomic Map