Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 7 - 8 | 856 | 69 | |
| GenBank mRNA | AF461761.1 | - | 7 - 8 | 755 | 69 | |
| AK051183.1 | - | 7 - 8 | 856 | 69 | ||
| AK090176.1 | - | 7 - 8 | 765 | 69 | ||
| AK166006.1 | - | 7 - 8 | 666 | 69 | ||
| BC029033.1 | - | 4 - 5 | 298 | 69 | ||
| BC033452.1 | - | 7 - 8 | 755 | 3156622 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a5
Genomic Map