| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300017290.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017290.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017290.mm.1 |
TC1300002540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017291.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017291.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017291.mm.1 |
TC1300002540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017289.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017289.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017289.mm.1 |
TC1300002540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017292.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017292.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017292.mm.1 |
TC1300002540.mm.1 |
MTA TC |