| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300007258.mm.1 |
NM_183115.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007258.mm.1 |
NM_183115.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007258.mm.1 |
TC1300001140.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007260.mm.1 |
NM_001168386.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007260.mm.1 |
NM_001168386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007260.mm.1 |
NM_183115.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007260.mm.1 |
NM_183115.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007260.mm.1 |
TC1300001140.mm.1 |
MTA TC |