Assay Details
| Gene Symbol: Naip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126182.2 | NP_001119654.1 | 2 - 3 | 139 | 76 | |
| NM_010872.3 | NP_035002.2 | 2 - 3 | 210 | 76 | ||
| GenBank mRNA | AF102871.1 | - | 169 | 76 | ||
| AF135489.1 | - | 2 - 3 | 180 | 76 | ||
| AF135490.1 | - | 2 - 3 | 109 | 76 | ||
| AF381770.1 | - | 2 - 3 | 109 | 76 | ||
| AY147000.1 | - | 2 - 3 | 109 | 76 | ||
| AY147001.1 | - | 2 - 3 | 109 | 76 | ||
| AY147004.1 | - | 1 - 2 | 42 | 76 | ||
| AY147005.1 | - | 1 - 2 | 44 | 76 | ||
| BC116626.1 | - | 1 - 2 | 49 | 40086453 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF102871.1, AF135489.1, AF135490.1, AF381770.1, AY147000.1, AY147001.1, AY147004.1, AY147005.1, BC116626.1, NM_001126182.2, NM_010872.3 transcripts.
Set Membership: |
|

Naip2
Genomic Map