Assay Details
| Gene Symbol: Gtf2h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145720.1 | - | 1 - 2 | 129 | 109 | |
| AK154739.1 | - | 1 - 2 | 123 | 109 | ||
| BC016231.1 | - | 1 - 2 | 104 | 4488798 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC016231.1 transcripts.
Set Membership: |
|

Gtf2h2
Genomic Map