Assay Details
| Gene Symbol: Gtf2h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022011.4 | NP_071294.3 | 3 - 4 | 326 | 141 | |
| GenBank mRNA | AB221674.1 | - | 2 - 3 | 140 | 141 | |
| AK078297.1 | - | 3 - 4 | 297 | 141 | ||
| AK145720.1 | - | 3 - 4 | 306 | 141 | ||
| AK154739.1 | - | 3 - 4 | 300 | 141 | ||
| AK158705.1 | - | 3 - 4 | 270 | 141 | ||
| BC016231.1 | - | 3 - 4 | 281 | 4488798 | 141 | |
| BC053382.1 | - | 3 - 4 | 290 | 30047823 | 141 | |
| BY277671.1 | - | 3 - 4 | 326 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2h2
Genomic Map