Assay Details
| Gene Symbol: Vcan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081249.1 | NP_001074718.1 | 12 - 13 | 10051 | 78 | |
| NM_001134474.1 | NP_001127946.1 | 11 - 12 | 4834 | 78 | ||
| NM_001134475.1 | NP_001127947.1 | 10 - 11 | 1954 | 78 | ||
| NM_019389.2 | NP_062262.2 | 11 - 12 | 7171 | 78 | ||
| GenBank mRNA | AK045314.1 | - | 5 - 6 | 3506 | 78 | |
| D16263.1 | - | 10 - 11 | 6743 | 78 | ||
| D28599.1 | - | 12 - 13 | 9795 | 78 | ||
| D32040.1 | - | 10 - 11 | 1686 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vcan
Genomic Map