Assay Details
| Gene Symbol: Xrcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028012.4 | NP_082288.1 | 6 - 7 | 849 | 92 | |
| GenBank mRNA | AB055154.1 | - | 6 - 7 | 757 | 92 | |
| AK009951.1 | - | 6 - 7 | 766 | 92 | ||
| AK053612.1 | - | 765 | 92 | |||
| AK088281.1 | - | 6 - 7 | 765 | 92 | ||
| BC025538.1 | - | 6 - 7 | 793 | 5257949 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xrcc4
Genomic Map