| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300016975.mm.1 |
NM_008634.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016975.mm.1 |
NM_008634.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016975.mm.1 |
TC1300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016976.mm.1 |
NM_008634.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016976.mm.1 |
NM_008634.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016976.mm.1 |
TC1300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016974.mm.1 |
NM_008634.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016974.mm.1 |
NM_008634.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016974.mm.1 |
TC1300002520.mm.1 |
MTA TC |