Assay Details
| Gene Symbol: Map1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008634.2 | NP_032660.2 | 3 - 4 | 488 | 72 | |
| GenBank mRNA | AK157749.1 | - | 1 - 2 | 260 | 72 | |
| AK158307.1 | - | 3 - 4 | 447 | 72 | ||
| AK158442.1 | - | 2 - 3 | 533 | 72 | ||
| BC019183.1 | - | 3 - 4 | 420 | 72 | ||
| BC037105.1 | - | 3 - 4 | 433 | 72 | ||
| BC046287.1 | - | 3 - 4 | 409 | 72 | ||
| BC052016.1 | - | 3 - 4 | 409 | 72 | ||
| BC078457.1 | - | 3 - 4 | 414 | 72 | ||
| BC138033.1 | - | 3 - 4 | 418 | 8861052 | 72 | |
| BC138034.1 | - | 3 - 4 | 418 | 8861053 | 72 | |
| X51396.1 | - | 3 - 4 | 426 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK157749.1 transcripts.
Set Membership: |
|

Map1b
Genomic Map