Assay Details
| Gene Symbol: Map1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008634.2 | NP_032660.2 | 4 - 5 | 631 | 104 | |
| GenBank mRNA | AK143169.1 | - | 2 - 3 | 365 | 104 | |
| AK157749.1 | - | 2 - 3 | 403 | 104 | ||
| AK158307.1 | - | 4 - 5 | 590 | 104 | ||
| BC019183.1 | - | 4 - 5 | 563 | 104 | ||
| BC037105.1 | - | 4 - 5 | 576 | 104 | ||
| BC046287.1 | - | 4 - 5 | 552 | 104 | ||
| BC052016.1 | - | 4 - 5 | 552 | 104 | ||
| BC078457.1 | - | 4 - 5 | 557 | 104 | ||
| BC138033.1 | - | 4 - 5 | 561 | 8861052 | 104 | |
| BC138034.1 | - | 4 - 5 | 561 | 8861053 | 104 | |
| X51396.1 | - | 4 - 5 | 569 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143169.1 transcripts.
Set Membership: |
|

Map1b
Genomic Map