Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 18 - 19 | 1787 | 83 | |
| GenBank mRNA | AK031041.1 | - | 18 - 19 | 1790 | 83 | |
| AK142282.1 | - | 18 - 19 | 1473 | 83 | ||
| AK164135.1 | - | 19 - 20 | 1645 | 83 | ||
| AK169439.1 | - | 2 - 3 | 1209 | 83 | ||
| BC052456.1 | - | 18 - 19 | 1484 | 6830607 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fcho2
Genomic Map