Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 4 - 5 | 689 | 81 | |
| GenBank mRNA | AK031041.1 | - | 4 - 5 | 692 | 81 | |
| AK142282.1 | - | 4 - 5 | 375 | 81 | ||
| AK146566.1 | - | 4 - 5 | 423 | 81 | ||
| AK164135.1 | - | 4 - 5 | 435 | 81 | ||
| BC019933.1 | - | 5 - 6 | 425 | 81 | ||
| BC047122.1 | - | 5 - 6 | 425 | 81 | ||
| BC052456.1 | - | 4 - 5 | 386 | 6830607 | 81 | |
| BC053718.1 | - | 4 - 5 | 405 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164135.1 transcripts.
Set Membership: |
|

Fcho2
Genomic Map