Assay Details
Gene Symbol: Tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025606.1 | NP_001020777.1 | 2 - 3 | 740 | 137 | |
GenBank mRNA | BC099490.1 | - | 2 - 3 | 740 | 30522938 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259031 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259034 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259035 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P799815 | BC099490.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P799815 | NM_001025606.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P799815 | BC099490.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P799815 | NM_001025606.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164576 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41794 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115046 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587662 | AK049391.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940926 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114329 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5763 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P799815 | BC099490.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P799815 | NM_001025606.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P799815 | BC099490.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P799815 | NM_001025606.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259026 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259035 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259030 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259052 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259053 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259060 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259025 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259062 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259065 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259063 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259055 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259040 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259057 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259014 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259027 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259050 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259058 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259022 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259045 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259028 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259039 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259066 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259044 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29259013 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07968060 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02756142 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07968069 | - | - |
Whole Mouse Genome G4122A | A_52_P799815 | BC099490.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P799815 | NM_001025606.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P799815 | BC099490.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P799815 | NM_001025606.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07968069 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10650071 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10650065 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10650070 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10650080 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10650062 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259031 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29259034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29259025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29258999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258976 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259026 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258943 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259075 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258947 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10542232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10926899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10758960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11010799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11049130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258990 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10523787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10475859 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259068 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10669660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11049132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10523788 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10574515 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10669665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258940 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10669667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10523791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10650063 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259018 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258992 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259008 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259016 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259030 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10447059 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10881934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259078 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259070 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10574513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10650082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259020 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258933 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259064 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259025 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259001 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10574514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10650071 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10475855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258979 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259027 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11049131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29258982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259065 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29259079 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10411460 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10411461 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10411462 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10411463 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456284_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10411459 | - | - |
Mouse Genome 430 2.0 Array | 1456284_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456284_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380863_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380863_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300016824.mm.1 | NM_001025606.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016824.mm.1 | NM_001025606.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016824.mm.1 | TC1300002506.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300016825.mm.1 | NM_001025606.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016825.mm.1 | NM_001025606.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016825.mm.1 | TC1300002506.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300016828.mm.1 | NM_001025606.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016828.mm.1 | NM_001025606.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016828.mm.1 | TC1300002506.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300016823.mm.1 | NM_001025606.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016823.mm.1 | NM_001025606.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016823.mm.1 | TC1300002506.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300016826.mm.1 | NM_001025606.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016826.mm.1 | NM_001025606.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300016826.mm.1 | TC1300002506.mm.1 | MTA TC |
Mouse Expression Set 430 | 1456284_PM_at | - | - |
Mouse Expression Set 430 | 1456284_at | - | - |
Murine Genome U74v2 Set | 165635_PM_i_at | - | - |
Murine Genome U74v2 Set | 165635_i_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456284_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456284_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 473229 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_2987487 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2987487 | - | - |
