Assay Details
| Gene Symbol: Tnpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048267.1 | NP_001041732.1 | 22 - 23 | 2699 | 98 | |
| NM_178716.3 | NP_848831.2 | 22 - 23 | 2705 | 98 | ||
| GenBank mRNA | AK029953.1 | - | 22 - 23 | 2706 | 98 | |
| AK030580.1 | - | 22 - 23 | 2665 | 98 | ||
| AK045132.1 | - | 22 - 23 | 2699 | 98 | ||
| AK163279.1 | - | 5 - 6 | 368 | 98 | ||
| BC031571.1 | - | 14 - 15 | 1748 | 98 | ||
| BC055372.1 | - | 22 - 23 | 2606 | 30059046 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tnpo1
Genomic Map