Assay Details
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027558.1 | NP_081834.1 | 1 - 2 | 456 | 61 | |
GenBank mRNA | AK014543.1 | - | 1 - 2 | 355 | 61 | |
AK140064.1 | - | 1 - 2 | 436 | 61 | ||
AX775753.1 | - | 1 - 2 | 412 | 61 | ||
BC044759.2 | - | 1 - 2 | 397 | 61 | ||
CD363842.1 | - | 1 - 2 | 455 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | AX775753.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | BC044759.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | AK140064.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | NM_027558.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | AK014543.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | AX775753.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | BC044759.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | AK140064.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | NM_027558.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413916 | AK014543.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372986 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | AK035542.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | AK083256.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | AK150971.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | AK151267.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | AK162081.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | BC054112.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | BC080290.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471272 | NM_001003948.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6753 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69537 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724679 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883520 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415784 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185177 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110487 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37809 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154913 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134189 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | AX775753.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | BC044759.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | AK140064.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | NM_027558.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | AK014543.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | AX775753.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | BC044759.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | AK140064.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | NM_027558.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413916 | AK014543.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134189 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154913 | - | - |
Whole Mouse Genome G4122A | A_52_P335326 | AX775753.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335326 | BC044759.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335326 | AK140064.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335326 | AK165560.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335326 | NM_027558.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | AX775753.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | BC044759.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | AK140064.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | NM_027558.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | AK014543.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | AX775753.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | BC044759.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | AK140064.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | NM_027558.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P413916 | AK014543.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154913 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082925 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082939 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082843 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082841 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02545389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02936185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02813134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02667618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082834 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082937 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02799734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082993 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082996 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083056 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02372345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082941 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02799737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082999 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02813137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03118862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03176626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083014 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02545395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083021 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22083039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22082960 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10497966 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10497967 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10497965 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452882_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428714_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10497964 | - | - |
Mouse Genome 430 2.0 Array | 1428714_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1428714_at | - | - |
Mouse Genome 430 2.0 Array | 1452882_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452882_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70804_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70804_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300014644.mm.1 | NM_027558.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300014644.mm.1 | NM_027558.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300014644.mm.1 | TC0300001909.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300014646.mm.1 | NM_027558.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300014646.mm.1 | NM_027558.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300014646.mm.1 | TC0300001909.mm.1 | MTA TC |
Mouse Expression Set 430 | 1428714_PM_at | - | - |
Mouse Expression Set 430 | 1428714_at | - | - |
Mouse Expression Set 430 | 1452882_PM_at | - | - |
Mouse Expression Set 430 | 1452882_at | - | - |
Murine Genome U74v2 Set | 161447_PM_f_at | - | - |
Murine Genome U74v2 Set | 94801_PM_at | - | - |
Murine Genome U74v2 Set | 94801_at | - | - |
Murine Genome U74v2 Set | 161447_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1428714_at | - | - |
Mouse Genome 430A 2.0 Array | 1428714_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452882_at | - | - |
Mouse Genome 430A 2.0 Array | 1452882_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428714_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452882_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428714_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452882_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 375231 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 103520673 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2524166 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1220641 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2715141 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103520673 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103520673 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220641 | - | - |
