Assay Details
| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177723.4 | NP_808391.2 | 2 - 3 | 352 | 69 | |
| GenBank mRNA | AK037228.1 | - | 2 - 3 | 353 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vsig8
Genomic Map