Assay Details
| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177723.4 | NP_808391.2 | 4 - 5 | 782 | 73 | |
| NR_027644.1 | 3 - 4 | 501 | 73 | |||
| GenBank mRNA | AK037228.1 | - | 4 - 5 | 783 | 73 | |
| BC064106.1 | - | 3 - 4 | 501 | 1528324 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vsig8
Genomic Map