Assay Details
| Gene Symbol: Cep89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028120.2 | NP_082396.1 | 8 - 9 | 962 | 81 | |
| GenBank mRNA | AK012067.1 | - | 8 - 9 | 963 | 81 | |
| AK029107.1 | - | 8 - 9 | 993 | 81 | ||
| BC003981.1 | - | 8 - 9 | 1017 | 3490246 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cep89
Genomic Map