Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100010634.mm.1 |
NM_001002004.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010634.mm.1 |
NM_001002004.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010634.mm.1 |
TC1100001153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010630.mm.1 |
NM_001002004.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010630.mm.1 |
NM_001002004.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010630.mm.1 |
TC1100001153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010631.mm.1 |
NM_001002004.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010631.mm.1 |
NM_001002004.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010631.mm.1 |
TC1100001153.mm.1 |
MTA TC |