Assay Details
| Gene Symbol: Aggf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025630.3 | NP_079906.2 | 11 - 12 | 2005 | 146 | |
| GenBank mRNA | AK009533.1 | - | 7 - 8 | 968 | 146 | |
| AK132575.1 | - | 11 - 12 | 2006 | 146 | ||
| AY500995.1 | - | 11 - 12 | 1978 | 146 | ||
| BC052410.1 | - | 11 - 12 | 1982 | 5716440 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aggf1
Genomic Map