Assay Details
| Gene Symbol: Aggf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025630.3 | NP_079906.2 | 12 - 13 | 2133 | 77 | |
| GenBank mRNA | AK009533.1 | - | 8 - 9 | 1096 | 77 | |
| AY500995.1 | - | 12 - 13 | 2106 | 77 | ||
| BC027286.1 | - | 7 - 8 | 77 | |||
| BC052410.1 | - | 12 - 13 | 2110 | 5716440 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aggf1
Genomic Map