Assay Details
| Gene Symbol: Ttc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290500.1 | NP_001277429.1 | 2 - 3 | 377 | 56 | |
| NM_001290502.1 | NP_001277431.1 | 2 - 3 | 615 | 56 | ||
| NM_025978.4 | NP_080254.2 | 2 - 3 | 377 | 56 | ||
| NM_027619.4 | NP_081895.2 | 2 - 3 | 377 | 56 | ||
| GenBank mRNA | AK012246.1 | - | 2 - 3 | 339 | 56 | |
| AK015308.1 | - | 2 - 3 | 337 | 56 | ||
| AK016619.1 | - | 2 - 3 | 297 | 56 | ||
| AK031605.1 | - | 2 - 3 | 608 | 56 | ||
| AK035570.1 | - | 2 - 3 | 347 | 56 | ||
| AK041095.1 | - | 2 - 3 | 351 | 56 | ||
| AK044266.1 | - | 2 - 3 | 352 | 56 | ||
| AK134539.1 | - | 2 - 3 | 366 | 56 | ||
| AK163444.1 | - | 2 - 3 | 377 | 56 | ||
| AK173315.1 | - | 2 - 3 | 347 | 56 | ||
| BC021448.1 | - | 2 - 3 | 385 | 56 | ||
| BC024847.1 | - | 2 - 3 | 342 | 5355806 | 56 | |
| BC138212.1 | - | 2 - 3 | 339 | 8861231 | 56 | |
| BC145944.1 | - | 2 - 3 | 339 | 40131247 | 56 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ttc14
Genomic Map