Assay Details
| Gene Symbol: Sec16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159986.1 | NP_001153458.1 | 7 - 8 | 1026 | 69 | |
| NM_033354.3 | NP_203505.3 | 9 - 10 | 1273 | 69 | ||
| NR_027641.1 | 8 - 9 | 1105 | 69 | |||
| GenBank mRNA | AK132144.1 | - | 7 - 8 | 1050 | 69 | |
| AK173302.1 | - | 9 - 10 | 1254 | 69 | ||
| BC059194.1 | - | 8 - 9 | 1091 | 4222915 | 69 | |
More Information
Set Membership: |
|

Sec16b
Genomic Map