Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300014210.mm.1 |
NM_001205361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014210.mm.1 |
NM_001205361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014210.mm.1 |
NM_001205362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014210.mm.1 |
NM_001205362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014210.mm.1 |
NM_033623.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014210.mm.1 |
NM_033623.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014210.mm.1 |
TC0300001841.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300014211.mm.1 |
NM_001205361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014211.mm.1 |
NM_001205361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014211.mm.1 |
NM_001205362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014211.mm.1 |
NM_001205362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014211.mm.1 |
NM_033623.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014211.mm.1 |
NM_033623.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300014211.mm.1 |
TC0300001841.mm.1 |
MTA TC |