Assay Details
Gene Symbol: Tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177839.3 | NP_808507.2 | 18 - 19 | 4234 | 62 | |
GenBank mRNA | AJ580920.1 | - | 15 - 16 | 3361 | 62 | |
AK048033.1 | - | 11 - 12 | 2349 | 62 | ||
BC138335.1 | - | 18 - 19 | 4195 | 8861355 | 62 | |
BC138336.1 | - | 18 - 19 | 4195 | 8861356 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
