Assay Details
| Gene Symbol: Npas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013780.2 | NP_038808.2 | 10 - 11 | 1314 | 81 | |
| GenBank mRNA | AF173871.1 | - | 10 - 11 | 1314 | 81 | |
| BC120885.1 | - | 9 - 10 | 1230 | 81 | ||
| BC148199.2 | - | 9 - 10 | 1230 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npas3
Genomic Map