Assay Details
| Gene Symbol: Nubpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029760.2 | NP_084036.2 | 2 - 3 | 294 | 56 | |
| GenBank mRNA | AK010827.1 | - | 2 - 3 | 276 | 56 | |
| AK083643.1 | - | 2 - 3 | 284 | 56 | ||
| AK141257.1 | - | 2 - 3 | 258 | 56 | ||
| AK152377.1 | - | 2 - 3 | 295 | 56 | ||
| BC062175.1 | - | 2 - 3 | 275 | 30291107 | 56 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nubpl
Genomic Map