Assay Details
| Gene Symbol: Gm527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025605.1 | NP_001020776.1 | 3 - 4 | 892 | 84 | |
| GenBank mRNA | BC099503.1 | - | 3 - 4 | 892 | 30940052 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm527
Genomic Map