Assay Details
Gene Symbol: Fam179b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177805.3 | NP_808473.2 | 17 - 18 | 4986 | 102 | |
GenBank mRNA | AK148829.1 | - | 5 - 6 | 636 | 102 | |
AK172948.1 | - | 17 - 18 | 4914 | 102 | ||
BC062107.1 | - | 7 - 8 | 1018 | 102 | ||
BC157958.1 | - | 17 - 18 | 4788 | 9088131 | 102 | |
BC158008.1 | - | 18 - 19 | 4938 | 9007316 | 102 | |
BC172117.1 | - | 17 - 18 | 4788 | 9054403 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
