Assay Details
| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076732.1 | - | 2 - 3 | 76 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076732.1 transcripts.
Set Membership: |
|

Larp1b
Genomic Map