Assay Details
| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040399.1 | NP_001035489.1 | 8 - 9 | 943 | 127 | |
| GenBank mRNA | AK076732.1 | - | 8 - 9 | 911 | 127 | |
| AK160155.1 | - | 8 - 9 | 943 | 127 | ||
| AK163348.1 | - | 7 - 8 | 909 | 127 | ||
| BC025528.1 | - | 6 - 7 | 866 | 5324328 | 127 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp1b
Genomic Map