Assay Details
| Gene Symbol: Pla2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004701.2 | - | 6 - 7 | 726 | 121 | |
| AK144750.1 | - | 7 - 8 | 683 | 121 | ||
| BC003816.1 | - | 7 - 8 | 723 | 3485483 | 121 | |
| M72394.1 | - | 7 - 8 | 671 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g4a
Genomic Map