| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100010997.mm.1 |
NM_001145807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010997.mm.1 |
NM_001145807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010997.mm.1 |
NM_153539.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010997.mm.1 |
NM_153539.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010997.mm.1 |
TC0100001330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010988.mm.1 |
NM_153539.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010988.mm.1 |
NM_153539.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010988.mm.1 |
TC0100001330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010989.mm.1 |
NM_153539.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010989.mm.1 |
NM_153539.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010989.mm.1 |
TC0100001330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011002.mm.1 |
NM_001145807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011002.mm.1 |
NM_001145807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011002.mm.1 |
NM_153539.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011002.mm.1 |
NM_153539.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011002.mm.1 |
TC0100001330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011001.mm.1 |
NM_001145807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011001.mm.1 |
NM_001145807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011001.mm.1 |
NM_153539.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011001.mm.1 |
NM_153539.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011001.mm.1 |
TC0100001330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011000.mm.1 |
NM_001145807.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011000.mm.1 |
NM_001145807.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011000.mm.1 |
NM_153539.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011000.mm.1 |
NM_153539.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011000.mm.1 |
TC0100001330.mm.1 |
MTA TC |