Assay Details
| Gene Symbol: Trmt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026876.3 | NP_081152.2 | 4 - 5 | 742 | 54 | |
| GenBank mRNA | AK148524.1 | - | 4 - 5 | 849 | 54 | |
| BC061491.1 | - | 4 - 5 | 748 | 54 | ||
| BC064024.1 | - | 4 - 5 | 512 | 54 | ||
| BC110425.1 | - | 4 - 5 | 725 | 5369595 | 54 | |
| BC132299.1 | - | 4 - 5 | 600 | 40130576 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt1l
Genomic Map