Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100008405.mm.1 |
NM_001122675.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008405.mm.1 |
NM_001122675.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008405.mm.1 |
NM_001122676.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008405.mm.1 |
NM_001122676.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008405.mm.1 |
TC0100001016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008406.mm.1 |
NM_001122675.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008406.mm.1 |
NM_001122675.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008406.mm.1 |
NM_001122676.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008406.mm.1 |
NM_001122676.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008406.mm.1 |
TC0100001016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100008407.mm.1 |
NM_001122675.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008407.mm.1 |
NM_001122675.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008407.mm.1 |
NM_001122676.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008407.mm.1 |
NM_001122676.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100008407.mm.1 |
TC0100001016.mm.1 |
MTA TC |