Assay Details
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081386.1 | NP_001074855.1 | 6 - 7 | 1195 | 110 | |
GenBank mRNA | AK148396.1 | - | 6 - 7 | 1183 | 110 | |
BC157927.1 | - | 6 - 7 | 1186 | 9088100 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
