Assay Details
| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081386.1 | NP_001074855.1 | 9 - 10 | 1684 | 105 | |
| GenBank mRNA | BC157927.1 | - | 9 - 10 | 1675 | 9088100 | 105 |
More Information
Set Membership: |
|

Cdh19
Genomic Map