Assay Details
Gene Symbol: Ppp1r12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081307.1 | NP_001074776.1 | 4 - 5 | 89 | ||
GenBank mRNA | AK042847.1 | - | 4 - 5 | 89 | ||
AK046012.1 | - | 4 - 5 | 89 | |||
AK046167.1 | - | 4 - 5 | 89 | |||
AK080855.1 | - | 4 - 5 | 89 | |||
AK141375.1 | - | 4 - 5 | 89 | |||
BC145746.1 | - | 4 - 5 | 769 | 40131049 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
