Assay Details
| Gene Symbol: Nr5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159769.2 | NP_001153241.1 | 3 - 4 | 337 | 147 | |
| NM_030676.3 | NP_109601.1 | 3 - 4 | 422 | 147 | ||
| GenBank mRNA | AB221629.1 | - | 3 - 4 | 264 | 147 | |
| AK054487.1 | - | 1 - 2 | 841 | 147 | ||
| BB652042.1 | - | 3 - 4 | 308 | 147 | ||
| BC137845.1 | - | 3 - 4 | 301 | 8860864 | 147 | |
| DQ133942.1 | - | 3 - 4 | 337 | 147 | ||
| M81385.1 | - | 3 - 4 | 422 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the DQ133942.1,NM_001159769.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nr5a2
Genomic Map