Assay Details
| Gene Symbol: Srgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081011.2 | NP_001074480.2 | 17 - 18 | 2589 | 83 | |
| GenBank mRNA | AK132220.1 | - | 3 - 4 | 262 | 83 | |
| AK161932.1 | - | 6 - 7 | 493 | 83 | ||
| BC030457.1 | - | 16 - 17 | 1896 | 83 | ||
| BC151081.1 | - | 16 - 17 | 2207 | 9087994 | 83 | |
| BC151082.1 | - | 16 - 17 | 2207 | 9087995 | 83 | |
| BC158055.1 | - | 16 - 17 | 2205 | 9088163 | 83 | |
| BC172152.1 | - | 16 - 17 | 2207 | 9054822 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srgap2
Genomic Map