Assay Details
| Gene Symbol: Rgsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243223.1 | NP_001230152.1 | 15 - 16 | 2689 | 91 | |
| GenBank mRNA | AK029630.1 | - | 6 - 7 | 796 | 91 | |
| AK161463.1 | - | 8 - 9 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rgsl1
Genomic Map